Cyclopentyltriphenylphosphonium bromide 98%

Supplier: Thermo Scientific Chemicals
Warning

A14647-09 A14647-18
CAAAA14647-09EA 84.53 CAD
CAAAA14647-09 CAAAA14647-18 CAAAA14647-30
Cyclopentyltriphenylphosphonium bromide 98%
Cyclopentyltriphenylphosphonium bromide

Purity: 98% CAS No.: 7333-52-0 Molecular Formula: C23H24BrP Formula Weight: 411.33 Melting Point: 262-266° Beilstein Reference: 3580193

Formula: C₂₃H₂₄BrP
Melting Pt: 262…266 °C
Storage Temperature: Ambient
MDL Number: MFCD00051880
CAS Number: 7333-52-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR