4-Isopropylaniline 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: p-Cumidine, 4-Aminocumene

A14646-14 A14646-22
CAAAA14646-14EA 94.96 CAD
CAAAA14646-14 CAAAA14646-22 CAAAA14646-36
4-Isopropylaniline 99%
4-Isopropylaniline

Purity: 99% CAS No.: 99-88-7 Molecular Formula: C9H13N Formula Weight: 135.21 Flash Point: 92°(197°F) Refractive Index: 1.5430 Melting Point: -63° Boiling Point: 226-228° Beilstein Reference: 1930544 Merck Reference: 14,2620 MDL No.: MFCD00007900

Formula: C₉H₁₃N
MW: 135.21 g/mol
Boiling Pt: 226
Melting Pt: -63
Density: 0.989
Flash Pt: 92 °C (197 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007900
CAS Number: 99-88-7
EINECS: 202-797-2
UN: 2735
ADR: 8 III
Merck Index: 14,2620

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR