2,4-Difluorobenzenesulfonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

A14639-22 A14639-06 A14639-14
CAAAAA14639-22EA 1202.38 CAD
CAAAAA14639-22 CAAAA14639-06 CAAAA14639-14
2,4-Difluorobenzenesulfonyl chloride 98%
2,4-Difluorobenzenesulphonyl chloride

Purity: 98% CAS No.: 13918-92-8 Molecular Formula: C6H3ClF2O2S Formula Weight: 212.60 Flash Point: 110°(230°F) Refractive Index: 1.5187 Boiling Point: 236-237° Beilstein Reference: 2970373 UN No.: 3265 MDL No.: MFCD00042481

Formula: C₆H₃ClF₂O₂S
Boiling Pt: 236…237 °C
Density: 1.581
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042481
CAS Number: 13918-92-8
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR