Thiosemicarbazide 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14630-0E
A14630-22
A14630-36
CAAAA14630-0EEA
595.66
CAD
CAAAA14630-0E
CAAAA14630-22
CAAAA14630-36
Thiosemicarbazide 98%
Thiosemicarbazide
Used in TLC to stain alpha-keto acids
Purity: 99%
CAS No.: 79-19-6
Molecular Formula: CH5N3S
Formula Weight: 91.14
Odor: Odorless
Melting Point: ca 180° dec.
Beilstein Reference: 506320
Merck Reference: 14,9361
UN No.: 2811
MDL No.: MFCD00007620
Formula:
CH₅N₃S Melting Pt: ∼ 180 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00007620 CAS Number: 79-19-6 EINECS: 201-184-7 UN: 2811 ADR: 6.1,III Merck Index: 14,9361 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...