Thiophene-3-carbaldehyde 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14628-09
A14628-18
CAAAA14628-09EA
152
CAD
CAAAA14628-09
CAAAA14628-18
CAAAA14628-30
Thiophene-3-carbaldehyde 96%
Thiophene-3-carbaldehyde
Purity: 96%
CAS No.: 498-62-4
Molecular Formula: C5H4OS
Formula Weight: 112.15
Storage Temperature: 2°C to 8°C
Flash Point: 64°(147°F)
Refractive Index: 1.5840
Boiling Point: 194-196°
Beilstein Reference: 105889
MDL No.: MFCD00005466
Formula:
C₅H₄OS Boiling Pt: 194…196 °C Melting Pt: –30 °C Density: 1.28 Flash Pt: 64 °C(147 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00005466 CAS Number: 498-62-4 EINECS: 207-865-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...