2',6'-Dihydroxyacetophenone 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(2,6-Dihydroxyphenyl)ethan-1-one, 1-(2,6-Dihydroxyphenyl)ethanone, 2,6-Dihydroxyacetophenone

A14627-06 A14627-14 A14627-22
CAAAA14627-06EA 53.56 CAD
CAAAA14627-06 CAAAA14627-14 CAAAA14627-22
2',6'-Dihydroxyacetophenone 98+%
2',6'-Dihydroxyacetophenone

Purity: 98+% CAS No.: 699-83-2 Molecular Formula: C8H8O3 Formula Weight: 152.15 Melting Point: 156-159° Beilstein Reference: 1366061

Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 156…159 °C
Storage Temperature: Ambient
MDL Number: MFCD00002270
CAS Number: 699-83-2
EINECS: 211-833-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR