1,2-Diphenylethane 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: Dibenzyl, Bibenzyl

A14617-14 A14617-22 A14617-36
CAAAA14617-14EA 95.59 CAD
CAAAA14617-14 CAAAA14617-22 CAAAA14617-36
1,2-Diphenylethane 98+%
1,2-Diphenylethane

MDL: MFCD00004796 Beilstein: 508068 Soluble in ether, chloroform


Purity: 98+% CAS No.: 103-29-7 Molecular Formula: C6H5CH2CH2C6H5 Formula Weight: 182.27 Flash Point: 110°(230°F) Melting Point: 50-53° Boiling Point: 284° Solubility: Soluble in ether, chloroform Beilstein Reference: 508068 Merck Reference: 14,1198 MDL No.: MFCD00004796

Formula: (C₆H₅)CH₂CH₂(C₆H₅)
Boiling Pt: 284 °C
Melting Pt: 50…53 °C
Density: 1.013
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004796
CAS Number: 103-29-7
EINECS: 203-096-4
Merck Index: 14,1198

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR