Sodium trifluoroacetate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Sodium 2,2,2-trifluoroacetate
A14613-14
A14613-22
A14613-36
CAAAA14613-14EA
61.62
CAD
CAAAA14613-14
CAAAA14613-22
CAAAA14613-36
Sodium trifluoroacetate 98%
Sodium trifluoroacetate
Purity: 98%
CAS No.: 2923-18-4
Molecular Formula: CF3CO2Na
Formula Weight: 136.00
Melting Point: ca 207° dec.
Beilstein Reference: 3597949
Formula:
C₂F₃NaO₂ MW: 136.01 g/mol Melting Pt: ∼ 207 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00013217 CAS Number: 2923-18-4 EINECS: 220-879-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...