2,4,6-Trichlorobenzonitrile 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14609-03
A14609-06
A14609-14
CAAAA14609-03EA
53.25
CAD
CAAAA14609-03
CAAAA14609-06
CAAAA14609-14
2,4,6-Trichlorobenzonitrile 97+%
2,4,6-Trichlorobenzonitrile
Purity: 97+%
CAS No.: 6575-05-9
Molecular Formula: C7H2Cl3N
Formula Weight: 206.46
Melting Point: 80-82°
UN No.: 3439
MDL No.: MFCD00052711
Formula:
C₇H₂Cl₃N Melting Pt: 80…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052711 CAS Number: 6575-05-9 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...