4,4'-Dimethoxybenzhydrol 98+%
Supplier: Thermo Scientific Chemicals
A14604-09
A14604-18
CAAAA14604-09EA
51.98
CAD
CAAAA14604-09
CAAAA14604-18
4,4'-Dimethoxybenzhydrol 98+%
4,4'-Dimethoxybenzhydrol
Purity: 98+%
CAS No.: 728-87-0
Molecular Formula: C15H16O3
Formula Weight: 244.29
Melting Point: 69-71°
Beilstein Reference: 748532
Formula:
C₁₅H₁₆O₃ Boiling Pt: 362 °C (1013 hPa) Melting Pt: 69…71 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008410 CAS Number: 728-87-0 EINECS: 211-975-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...