Sarcosine 98%
Supplier: Thermo Scientific Chemicals
A14594-0E
A14594-22
A14594-36
CAAAAA14594-0EEA
756.82
CAD
CAAAAA14594-0E
CAAAA14594-22
CAAAA14594-36
Sarcosine 98%
Sarcosine
A type 1 glycine transporter inhibitor
Purity: 98%
CAS No.: 107-97-1
Molecular Formula: CH3NHCH2CO2H
Formula Weight: 89.09
Odor: Odorless
pH: 6.1
Melting Point: ca 208° dec.
Beilstein Reference: 1699442
Merck Reference: 14,8373
MDL No.: MFCD00004279
Formula:
C₃H₇NO₂ Boiling Pt: 185 °C (1013 hPa) Melting Pt: ∼ 208 °C (dec.) Density: 1.301 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004279 CAS Number: 107-97-1 EINECS: 203-538-6 Merck Index: 14,8373 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...