2-Acetyl-1H-pyrrole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl-2-pyrrolylketon, Methyl 2-pyrrolyl ketone, 1-(1H-Pyrrol-2-yl)ethanone, 1-(1H-Pyrrol-2-yl)ethan-1-one, 2-Acetylpyrrole

A14593-06 A14593-14
CAAAA14593-03EA 21.63 CAD
CAAAA14593-03 CAAAA14593-06 CAAAA14593-14
2-Acetyl-1H-pyrrole 98%
2-Acetyl-1H-pyrrole

Purity: 98% CAS No.: 1072-83-9 Molecular Formula: C6H7NO Formula Weight: 109.13 Odor: Characteristic Melting Point: 88-92° Boiling Point: 220° Beilstein Reference: 1882 MDL No.: MFCD00005220

Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 220 °C
Melting Pt: 88…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00005220
CAS Number: 1072-83-9
EINECS: 214-016-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR