2,3-Dimethoxybenzoic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14592-09
A14592-18
CAAAA14592-09EA
73.79
CAD
CAAAA14592-09
CAAAA14592-18
CAAAA14592-30
2,3-Dimethoxybenzoic acid 99%
2,3-Dimethoxybenzoic acid
Purity: 99%
CAS No.: 1521-38-6
Molecular Formula: C9H10O4
Formula Weight: 182.18
Odor: Odorless
Melting Point: 121-124°
Beilstein Reference: 2210858
MDL No.: MFCD00002432
Formula:
C₉H₁₀O₄ Boiling Pt: 314 °C (1013 hPa) Melting Pt: 121…124 °C Density: 1,387 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002432 CAS Number: 1521-38-6 EINECS: 216-188-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...