Zinc carbonate basic (>58.0% Zn) 97% Zn >58.0%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14590-0B
A14590-30
CAAAA14590-0BEA
181.7
CAD
CAAAA14590-0B
CAAAA14590-30
Zinc carbonate basic (>58.0% Zn) 97% Zn >58.0%
Zinc carbonate basic
Purity: 97%
CAS No.: 5263-02-5
Molecular Formula: Zn5(CO3)2(OH)6
Formula Weight: 548.97
MDL No.: MFCD00151660
Formula:
C₂H₆O₁₂Zn₅ Melting Pt: > 140 °C Density: 3.5 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011519 CAS Number: 5263-02-5 EINECS: 226-076-7 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...