4-Pyridone 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Hydroxypyridine, γ-Pyridone, 4-Pyridinol, Pyridin-4-ol, 1,4-Dihydro-4-Pyridinone

A14586-14 A14586-22 A14586-36
CAAAA14586-14EA 100.49 CAD
CAAAA14586-14 CAAAA14586-22 CAAAA14586-36
4-Pyridone 97%
4-Pyridone

Purity: 97% CAS No.: 626-64-2 Molecular Formula: C5H5NO Formula Weight: 95.10 Melting Point: 142-148° Boiling Point: 230-235°/12mm MDL No.: MFCD00040458

Formula: C₅H₅NO
MW: 95.1 g/mol
Boiling Pt: 230…235 °C (12 mmHg)
Melting Pt: 142…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00006419
CAS Number: 626-64-2
EINECS: 210-958-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR