2'-Hydroxy-4',6'-dimethoxyacetophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(2-Hydroxy-4,6-dimethoxyphenyl)ethan-1-one, Xanthoxylin

A14585-03 A14585-06
CAAAA14585-03EA 87.06 CAD
CAAAA14585-03 CAAAA14585-06
2'-Hydroxy-4',6'-dimethoxyacetophenone 98%
2'-Hydroxy-4',6'-dimethoxyacetophenone

Purity: 98% CAS No.: 90-24-4 Molecular Formula: C10H12O4 Formula Weight: 196.20 Melting Point: 80-83° Boiling Point: 185°/20mm Merck Reference: 14,10067 MDL No.: MFCD00017243

Formula: C₁₀H₁₂O₄
MW: 196.2
Boiling Pt: 185 °C (20 mmHg)
Melting Pt: 80…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00017243
CAS Number: 90-24-4
EINECS: 201-978-3
Merck Index: 14,10067

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR