6-Chloro-1-hexanol 97%

Supplier: Thermo Scientific Chemicals
Warning

A14582-14 A14582-22 A14582-36
CAAAA14582-14EA 100.01 CAD
CAAAA14582-14 CAAAA14582-22 CAAAA14582-36
6-Chloro-1-hexanol 97%
6-Chloro-1-hexanol

Purity: 97% CAS No.: 2009-83-8 Molecular Formula: Cl(CH2)6OH Formula Weight: 136.62 Flash Point: 98°(208°F) Refractive Index: 1.4560 Boiling Point: 109-111°/14mm Beilstein Reference: 1697307 MDL No.: MFCD00002984

Formula: C₆H₁₃ClO
Boiling Pt: 109…111 °C/14 mm
Density: 1.027
Flash Pt: 98 °C (208 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002984
CAS Number: 2009-83-8
EINECS: 217-925-2
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR