3-Bromoquinoline 98%

Supplier: Thermo Scientific Chemicals
Warning

A14573-22 A14573-09 A14573-18
CAAAAA14573-22EA 418.7 CAD
CAAAAA14573-22 CAAAA14573-09 CAAAA14573-18 CAAAA14573-30
3-Bromoquinoline 98%
3-Bromoquinoline

Purity: 98% CAS No.: 5332-24-1 Molecular Formula: C9H6BrN Formula Weight: 208.06 Flash Point: 110°(230°F) Refractive Index: 1.6640 Melting Point: 13-15° Boiling Point: 274-276° Beilstein Reference: 112939 MDL No.: MFCD00006767

Formula: C₉H₆BrN
MW: 208.06 g/mol
Boiling Pt: 274…276 °C
Melting Pt: 13…15 °C
Density: 1.533
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006767
CAS Number: 5332-24-1
EINECS: 226-237-1
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR