1,2,3-Trimethoxybenzene 98+%

Supplier: Thermo Scientific Chemicals

A14572-14 A14572-22 A14572-36
CAAAA14572-14EA 57.04 CAD
CAAAA14572-14 CAAAA14572-22 CAAAA14572-36
1,2,3-Trimethoxybenzene 98+%
1,2,3-Trimethoxybenzene

Purity: 98+% CAS No.: 634-36-6 Molecular Formula: C9H12O3 Formula Weight: 168.19 Flash Point: 112°(233°F) Melting Point: 43-45° Boiling Point: 241° Beilstein Reference: 1910422 MDL No.: MFCD00008358

Formula: C₉H₁₂O₃
MW: 168.19 g/mol
Boiling Pt: 241
Melting Pt: 43…45 °C
Density: 1.112
Flash Pt: 112 °C (233 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008358
CAS Number: 634-36-6
EINECS: 211-207-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR