3-(4-Hydroxyphenyl)propionic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14567-30
A14567-09
A14567-18
CAAAAA14567-30EA
692.04
CAD
CAAAAA14567-30
CAAAA14567-09
CAAAA14567-18
3-(4-Hydroxyphenyl)propionic acid 99%
3-(4-Hydroxyphenyl)propionic acid
Purity: 99%
CAS No.: 501-97-3
Molecular Formula: C9H10O3
Formula Weight: 166.18
Melting Point: 126-131°
Beilstein Reference: 2209841
Formula:
C₉H₁₀O₃ Boiling Pt: 208…210 °C (14 mmHg) Melting Pt: 126…131 °C Density: 1,336 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002778 CAS Number: 501-97-3 EINECS: 207-931-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...