2-Naphthol 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Hydroxynaphthalene, β-Naphthol, β-Hydroxynaphthalene

A14564-0B A14564-0I A14564-30
CAAAA14564-0BEA 109.81 CAD
CAAAA14564-0B CAAAA14564-0I CAAAA14564-30
2-Naphthol 98+%
2-Naphthol

Fluorescence indicator


Purity: 98+% CAS No.: 135-19-3 Molecular Formula: C10H8O Formula Weight: 144.17 Flash Point: 153°(307°F) Odor: Phenol-like pH: 5.77 Melting Point: 120-124° Boiling Point: 285-286° Beilstein Reference: 742134 Merck Reference: 14,6384 UN No.: 3077 MDL No.: MFCD00004067

Formula: C₁₀H₈O
Boiling Pt: 285…286 °C
Melting Pt: 120…124 °C
Density: 1.280
Flash Pt: 153 °C (307 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004067
CAS Number: 135-19-3
EINECS: 205-182-7
UN: 3077
ADR: 9,III
Merck Index: 14,6384

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR