Thymol 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Isopropyl-3-hydroxytoluene, 2-Isopropyl-5-methylphenol, 6-Isopropyl-m-cresol

A14563-30 A14563-22 A14563-36
CAAAAA14563-30EA 102.86 CAD
CAAAAA14563-30 CAAAA14563-22 CAAAA14563-36
Thymol 98+%
Thymol

Purity: 98+% CAS No.: 89-83-8 Molecular Formula: C10H14O Formula Weight: 150.22 Flash Point: 102°(215°F) pH: 5-7 Melting Point: 49-52° Boiling Point: 231-233° Beilstein Reference: 1907135 Merck Reference: 14,9399 UN No.: 2430 MDL No.: MFCD00002309

Formula: 2-[(CH₃)₂CH]C₆H₃-5-(CH₃)OH
MW: 150.22 g/mol
Boiling Pt: 233
Melting Pt: 49…52 °C
Density: 0.970
Flash Pt: 102 °C (215 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002309
CAS Number: 89-83-8
EINECS: 201-944-8
UN: 2430
ADR: 8 III
Merck Index: 14,9399
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b3268b34e160525798cab810087412f9b9a9db6&vid=7293cafb537e97291a403e856ca7fdb0223ba714&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR