2,6-Difluorobenzoic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14558-14
A14558-22
CAAAAA14558-36EA
2455.72
CAD
CAAAAA14558-36
CAAAA14558-14
CAAAA14558-22
2,6-Difluorobenzoic acid 98+%
2,6-Difluorobenzoic acid
Purity: 98+%
CAS No.: 385-00-2
Molecular Formula: C7H4F2O2
Formula Weight: 158.11
Odor: Odorless
Melting Point: 156-160°
Boiling Point: 72-77°/13mm
Beilstein Reference: 973774
MDL No.: MFCD00002411
Formula:
C₇H₄F₂O₂ Boiling Pt: 72…77 °C/13 mm Melting Pt: 156…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002411 CAS Number: 385-00-2 EINECS: 206-856-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...