3,5-Dinitro-o-toluic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14555-36
A14555-14
A14555-22
CAAAAA14555-36EA
259.12
CAD
CAAAAA14555-36
CAAAA14555-14
CAAAA14555-22
3,5-Dinitro-o-toluic acid 98%
3,5-Dinitro-o-toluic acid
Purity: 98%
CAS No.: 28169-46-2
Molecular Formula: C8H6N2O6
Formula Weight: 226.15
Melting Point: 203-207°
Beilstein Reference: 2622906
Formula:
C₈H₆N₂O₆ Melting Pt: 203…207 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007161 CAS Number: 28169-46-2 EINECS: 248-880-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...