Potassium formate, (max. 2% H₂O) 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14551-0B
A14551-0I
A14551-30
CAAAA14551-0BEA
184.86
CAD
CAAAA14551-0B
CAAAA14551-0I
CAAAA14551-30
Potassium formate, (max. 2% H₂O) 99%
Potassium formate
MDL: MFCD00013100
Beilstein: 3623272
Soluble in water
Purity: 99%
CAS No.: 590-29-4
Molecular Formula: HCO2K
Formula Weight: 84.12
Melting Point: 165-168°
Solubility: Soluble in water
Beilstein Reference: 3623272
Merck Reference: 14,7633
MDL No.: MFCD00013100
Formula:
CHKO₂ Melting Pt: 165…168 °C Density: 1.91 Storage Temperature: Ambient |
MDL Number:
MFCD00013100 CAS Number: 590-29-4 EINECS: 209-677-9 Merck Index: 14,7633 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...