Fumaroyl chloride 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14540-14
A14540-22
CAAAA14540-14EA
82.95
CAD
CAAAA14540-14
CAAAA14540-22
Fumaroyl chloride 95%
Fumaroyl chloride
Purity: 95%
CAS No.: 627-63-4
Molecular Formula: C4H2Cl2O2
Formula Weight: 152.97
Flash Point: 73°(163°F)
Refractive Index: 1.5000
Odor: Acrid
Melting Point: -2°
Boiling Point: 159-161°
Beilstein Reference: 1721342
UN No.: 1780
MDL No.: MFCD00000733
Formula:
C₄H₂Cl₂O₂ Boiling Pt: 159…161 °C Melting Pt: –2 °C Density: 1.413 Flash Pt: 73 °C (163 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000733 CAS Number: 627-63-4 EINECS: 211-005-4 UN: 1780 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...