2-Butyne-1,4-diol 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14539-0B
A14539-0I
A14539-30
CAAAA14539-0BEA
112.5
CAD
CAAAA14539-0B
CAAAA14539-0I
CAAAA14539-30
2-Butyne-1,4-diol 98+%
2-Butyne-1,4-diol
Purity: 98+%
CAS No.: 110-65-6
Molecular Formula: C4H6O2
Formula Weight: 86.09
Flash Point: 152°(305°F)
Melting Point: 54-58°
Boiling Point: 194°/100mm
Beilstein Reference: 1071237
UN No.: 2716
MDL No.: MFCD00002915
Formula:
C₄H₆O₂ Boiling Pt: 194 °C/100 mm Melting Pt: 54…58 °C Density: 1.1001 g/cm³ (20 °C) Flash Pt: 152 °C (305 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002915 CAS Number: 110-65-6 EINECS: 203-788-6 UN: 2716 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...