1-Phenyl-1,3-butandione 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-Phenylbutane-1,3-dione, 1-Benzoylacetone, Benzoylacetone, 1-Phenyl-1,3-butanedione

A14537-0B A14537-18 A14537-30
CAAAAA14537-0BEA 1140.76 CAD
CAAAAA14537-0B CAAAA14537-18 CAAAA14537-30
1-Phenyl-1,3-butandione 98+%
1-Phenyl-1,3-butandione

Purity: 98+% CAS No.: 93-91-4 Molecular Formula: C10H10O2 Formula Weight: 162.19 Melting Point: 55-60° Boiling Point: 260-262° MDL No.: MFCD00008786

Formula: C₆H₅C(O)CH₂C(O)CH₃
Boiling Pt: 260…262 °C
Melting Pt: 55…60 °C
Density: 1.090
Storage Temperature: Ambient
MDL Number: MFCD00008786
CAS Number: 93-91-4
EINECS: 202-286-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR