2,4-Dinitrobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A14536-06 A14536-14
CAAAA14536-06EA 44.71 CAD
CAAAA14536-06 CAAAA14536-14
2,4-Dinitrobenzoic acid 98%
2,4-Dinitrobenzoic acid

Purity: 98% CAS No.: 610-30-0 Molecular Formula: C7H4N2O6 Formula Weight: 212.12 Melting Point: 178-184° Beilstein Reference: 658650

Formula: C₇H₄N₂O₆
Melting Pt: 178…184 °C
Density: 1,672 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00017006
CAS Number: 610-30-0
EINECS: 210-219-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR