4-Chlorobenzonitrile 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14524-0B
A14524-18
A14524-30
CAAAA14524-0BEA
1350.9
CAD
CAAAA14524-0B
CAAAA14524-18
CAAAA14524-30
4-Chlorobenzonitrile 99%
4-Chlorobenzonitrile
Purity: 99%
CAS No.: 623-03-0
Molecular Formula: C7H4ClN
Formula Weight: 137.57
Flash Point: 108°(226°F)
Odor: Almond-like
Boiling Point: 222-224°
Beilstein Reference: 1072122
MDL No.: MFCD00001813
Formula:
C₇H₄ClN Boiling Pt: 222…224 °C Melting Pt: 90…94 °C Density: 1,200 Flash Pt: 108 °C(226 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001813 CAS Number: 623-03-0 EINECS: 210-765-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...