2-Bromobenzoyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Bromobenzene-1-carbonyl chloride, o-Bromobenzoyl chloride

A14520-30 A14520-09 A14520-18
CAAAAA14520-30EA 872.16 CAD
CAAAAA14520-30 CAAAA14520-09 CAAAA14520-18
2-Bromobenzoyl chloride 98%
2-Bromobenzoyl chloride

Purity: 98% CAS No.: 7154-66-7 Molecular Formula: C7H4BrClO Formula Weight: 219.47 Flash Point: 107°(224°F) Refractive Index: 1.5960 Melting Point: 10° Boiling Point: 245° Beilstein Reference: 508506 UN No.: 3265 MDL No.: MFCD00000655

Formula: C₇H₄BrClO
Boiling Pt: 245 °C
Melting Pt: 10 °C
Density: 1.674
Flash Pt: 107 °C (224 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000655
CAS Number: 7154-66-7
EINECS: 230-507-4
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR