N-(4-Bromobutyl)phthalimide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14517-06
A14517-14
A14517-22
CAAAA14517-06EA
88.01
CAD
CAAAA14517-06
CAAAA14517-14
CAAAA14517-22
N-(4-Bromobutyl)phthalimide 98%
N-(4-Bromobutyl)phthalimide
Purity: 98%
CAS No.: 5394-18-3
Molecular Formula: C12H12BrNO2
Formula Weight: 282.14
Odor: Odorless
Melting Point: 77-81°
Boiling Point: 165-170°/1mm
Beilstein Reference: 164119
MDL No.: MFCD00005905
Formula:
C₁₂H₁₂BrNO₂ Boiling Pt: 165…170 °C/1 mm Melting Pt: 77…81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005905 CAS Number: 5394-18-3 EINECS: 226-401-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...