2,6-Xylidine 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14512-22
A14512-36
A14512-0B
CAAAAA14512-22EA
50.54
CAD
CAAAAA14512-22
CAAAAA14512-36
CAAAA14512-0B
2,6-Xylidine 99%
2,6-Xylidine
Purity: 99%
CAS No.: 87-62-7
Molecular Formula: C8H11N
Formula Weight: 121.18
Flash Point: 91°(195°F)
Refractive Index: 1.5600
Odor: Characteristic
Melting Point: 11°
Boiling Point: 216°
Beilstein Reference: 636332
Merck Reference: 14,10084
UN No.: 1711
MDL No.: MFCD00007747
Formula:
C₈H₁₁N Boiling Pt: 216 °C Melting Pt: 11 °C Density: 0.981 Flash Pt: 91 °C (195 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007747 CAS Number: 87-62-7 EINECS: 201-758-7 UN: 1711 ADR: 6.1,II Merck Index: 14,10084 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...