2-Methyl-9,10-anthraquinone 97%
Supplier: Thermo Scientific Chemicals
A14502-0B
A14502-18
A14502-30
CAAAA14502-0BEA
477.16
CAD
CAAAA14502-0B
CAAAA14502-18
CAAAA14502-30
2-Methyl-9,10-anthraquinone 97%
2-Methyl-9,10-anthraquinone
Purity: 97%
CAS No.: 84-54-8
Molecular Formula: C15H10O2
Formula Weight: 222.24
Flash Point: 209°(408°F)
Melting Point: 170-174°
Beilstein Reference: 2050523
Merck Reference: 14,6021
MDL No.: MFCD00001235
Formula:
C₁₅H₁₀O₂ Boiling Pt: 236 - 238 °C (10 -rr) Melting Pt: 170…174 °C Density: 1.365 g/cm³ (20 °C) Flash Pt: 209 °C(408 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001235 CAS Number: 84-54-8 EINECS: 201-539-6 UN: 3077 ADR: 9,III Merck Index: 14,6021 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...