1-Propyl isothiocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14499-06
A14499-14
CAAAA14499-06EA
77.58
CAD
CAAAA14499-06
CAAAA14499-14
CAAAA14499-22
1-Propyl isothiocyanate 97%
1-Propyl isothiocyanate
Purity: 97%
CAS No.: 628-30-8
Molecular Formula: CH3(CH2)2NCS
Formula Weight: 101.17
Flash Point: 46°(114°F)
Refractive Index: 1.5040
Odor: Pungent
Boiling Point: 152-154°
Beilstein Reference: 1740199
UN No.: 2924
MDL No.: MFCD00004823
Formula:
C₄H₇NS MW: 101.17 Boiling Pt: 152…154 °C Density: 0.977 Flash Pt: 46 °C (114 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004823 CAS Number: 628-30-8 EINECS: 211-035-8 UN: 2924 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...