Diethyl oxalate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Oxalic acid diethyl ester

A14498-0C A14498-0E A14498-36
CAAAA14498-0CEA 521.4 CAD
CAAAA14498-0C CAAAA14498-0E CAAAA14498-36
Diethyl oxalate 99%
Diethyl oxalate

Purity: 99% CAS No.: 95-92-1 Molecular Formula: C6H10O4 Formula Weight: 146.14 Flash Point: 75°(167°F) Refractive Index: 1.4100 Melting Point: -41° Boiling Point: 184-186° Beilstein Reference: 606350 Merck Reference: 14,3125 UN No.: 2525 MDL No.: MFCD00009119

Formula: C₆H₁₀O₄
MW: 146.14 g/mol
Boiling Pt: 184…186 °C
Melting Pt: –41 °C
Density: 1.077
Flash Pt: 75 °C (167 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009119
CAS Number: 95-92-1
EINECS: 202-464-1
UN: 2525
ADR: 6.1,III
Merck Index: 14,3125

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR