1-Bromo-3-methylbutane 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14492-0B
A14492-0I
A14492-30
CAAAAA14492-22EA
66.49
CAD
CAAAAA14492-22
CAAAA14492-0B
CAAAA14492-0I
CAAAA14492-30
1-Bromo-3-methylbutane 98%
1-Bromo-3-methylbutane
Purity: 98%
CAS No.: 107-82-4
Molecular Formula: C5H11Br
Formula Weight: 151.05
Flash Point: 22°(71°F)
Refractive Index: 1.4409
Melting Point: -112°
Boiling Point: 120-121°
Beilstein Reference: 773652
Merck Reference: 14,5114
UN No.: 2341
MDL No.: MFCD00000253
Formula:
C₅H₁₁Br Boiling Pt: 120…121 °C Melting Pt: –112 °C Density: 1.208 Flash Pt: 22 °C (71 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000253 CAS Number: 107-82-4 EINECS: 203-522-9 UN: 2341 ADR: 3,III Merck Index: 14,5114 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...