(±)-3-Octanol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14488-18
A14488-30
CAAAA14488-18EA
70.94
CAD
CAAAA14488-18
CAAAA14488-30
(±)-3-Octanol 97%
(±)-3-Octanol
Purity: 97%
CAS No.: 589-98-0
Molecular Formula: C8H18O
Formula Weight: 130.23
Flash Point: 68°(154°F)
Refractive Index: 1.4270
Odor: Pleasant
Melting Point: -45°
Boiling Point: 175-176°
Beilstein Reference: 1719310
MDL No.: MFCD00004590
Formula:
C₈H₁₈O Boiling Pt: 175…176 °C Melting Pt: –45 °C Density: 0.820 Flash Pt: 68 °C(154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004590 CAS Number: 589-98-0 EINECS: 209-667-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...