2,6-Dihydroxybenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A14485-36 A14485-14 A14485-22
CAAAAA14485-36EA 722.06 CAD
CAAAAA14485-36 CAAAA14485-14 CAAAA14485-22
2,6-Dihydroxybenzoic acid 98%
2,6-Dihydroxybenzoic acid

Purity: 98% CAS No.: 303-07-1 Molecular Formula: C7H6O4 Formula Weight: 154.12 Melting Point: ca 164° dec. Beilstein Reference: 2209755

Formula: C₇H₆O₄
MW: 154.12 g/mol
Boiling Pt: 339
Melting Pt: ∼ 164 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00002462
CAS Number: 303-07-1
EINECS: 206-134-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR