Cyclobutanecarboxylic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14480-14
A14480-22
CAAAA14480-14EA
150.1
CAD
CAAAA14480-14
CAAAA14480-22
Cyclobutanecarboxylic acid 98+%
Cyclobutanecarboxylic acid
Purity: 98+%
CAS No.: 3721-95-7
Molecular Formula: C5H8O2
Formula Weight: 100.12
Flash Point: 83°(181°F)
Refractive Index: 1.4433
Odor: Stench
Melting Point: -20°
Boiling Point: 194-195°
Beilstein Reference: 1816777
UN No.: 3265
MDL No.: MFCD00001323
Formula:
C₅H₈O₂ Boiling Pt: 194…195 °C Melting Pt: –20 °C Density: 1.055 Flash Pt: 83 °C(181 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001323 CAS Number: 3721-95-7 EINECS: 223-072-7 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...