Pentafluoropropionic anhydride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14472-06
A14472-14
A14472-22
CAAAA14472-06EA
63.83
CAD
CAAAA14472-06
CAAAA14472-14
CAAAA14472-22
Pentafluoropropionic anhydride 98%
Pentafluoropropionic anhydride
Purity: 98%
CAS No.: 356-42-3
Molecular Formula: (CF3CF2CO)2O
Formula Weight: 310.04
Boiling Point: 73-74°
Beilstein Reference: 1806446
UN No.: 3265
MDL No.: MFCD00000429
Formula:
C₆F₁₀O₃ Boiling Pt: 73…74 °C Density: 1.589 Storage Temperature: Ambient |
MDL Number:
MFCD00000429 CAS Number: 356-42-3 EINECS: 206-604-2 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...