4'-Methylacetophenone 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl p-tolyl ketone, p-Methylacetophenone, 1-(p-Tolyl)ethanone
A14469-0B
A14469-18
A14469-30
CAAAA14469-0BEA
152.15
CAD
CAAAA14469-0B
CAAAA14469-18
CAAAA14469-30
4'-Methylacetophenone 96%
4'-Methylacetophenone
Purity: 96%
CAS No.: 122-00-9
Molecular Formula: C9H10O
Formula Weight: 134.18
Flash Point: 92°(197°F)
Refractive Index: 1.5340
Melting Point: -19°
Boiling Point: 224-226°
Beilstein Reference: 606053
MDL No.: MFCD00008751
Formula:
C₉H₁₀O MW: 134.18 g/mol Boiling Pt: 224…226 °C Melting Pt: –19 °C Density: 1.010 Flash Pt: 92 °C(197 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008751 CAS Number: 122-00-9 EINECS: 204-514-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...