4-Methoxyphenylboronic acid 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14462-03
A14462-06
A14462-14
CAAAA14462-03EA
59.41
CAD
CAAAA14462-03
CAAAA14462-06
CAAAA14462-14
4-Methoxyphenylboronic acid 97+%
4-Methoxyphenylboronic acid
Purity: 97+%
CAS No.: 5720-07-0
Molecular Formula: C7H9BO3
Formula Weight: 151.96
Melting Point: 202-208°
Beilstein Reference: 2936912
Formula:
C₇H₉BO₃ Melting Pt: 202…208 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039139 CAS Number: 5720-07-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...