2,6-Dimethylbenzonitrile 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14461-03
A14461-06
A14461-14
CAAAA14461-03EA
58.78
CAD
CAAAA14461-03
CAAAA14461-06
CAAAA14461-14
2,6-Dimethylbenzonitrile 97%
2,6-Dimethylbenzonitrile
Purity: 97%
CAS No.: 6575-13-9
Molecular Formula: C9H9N
Formula Weight: 131.18
Odor: Almond-like
Melting Point: 88-92°
Beilstein Reference: 1932523
MDL No.: MFCD00037166
Formula:
C₉H₉N Melting Pt: 88…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037166 CAS Number: 6575-13-9 EINECS: 229-503-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...