Trifluoroacetamide 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,2,2-Trifluoroacetamide

A14451-14 A14451-22 A14451-36
CAAAA14451-14EA 62.73 CAD
CAAAA14451-14 CAAAA14451-22 CAAAA14451-36
Trifluoroacetamide 97%
Trifluoroacetamide

Purity: 97% CAS No.: 354-38-1 Molecular Formula: CF3CONH2 Formula Weight: 113.04 Melting Point: 70-75° Boiling Point: 162-164° MDL No.: MFCD00008008

Formula: C₂H₂F₃NO
MW: 113.04 g/mol
Boiling Pt: 162…164 °C
Melting Pt: 70…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00008008
CAS Number: 354-38-1
EINECS: 206-559-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR