Phthalic acid 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14450-0B
A14450-0I
A14450-30
CAAAA14450-0BEA
178.54
CAD
CAAAA14450-0B
CAAAA14450-0I
CAAAA14450-30
Phthalic acid 99%
Phthalic acid
MDL: MFCD00002467
Beilstein: 608199
Soluble in water, alcohol and ether
Purity: 99%
CAS No.: 88-99-3
Molecular Formula: C8H6O4
Formula Weight: 166.13
Flash Point: 168°(334°F)
Odor: Odorless
Beilstein Reference: 608199
Merck Reference: 14,7371
MDL No.: MFCD00002467
Formula:
C₈H₆O₄ Boiling Pt: 315 - 317 °C (1013 hPa) Melting Pt: 205 °C (dec.) Density: 1.593 Flash Pt: 168 °C(334 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002467 CAS Number: 88-99-3 EINECS: 201-873-2 Merck Index: 14,7371 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...