Isophthalic acid 99%

Supplier: Thermo Scientific Chemicals

A14445-0B A14445-0I A14445-30
CAAAA14445-0BEA 53.09 CAD
CAAAA14445-0B CAAAA14445-0I CAAAA14445-30
Isophthalic acid 99%
Isophthalic acid

Purity: 99% CAS No.: 121-91-5 Molecular Formula: C8H6O4 Formula Weight: 166.13 Melting Point: 344-348° subl. Beilstein Reference: 1909332 Merck Reference: 14,5197 MDL No.: MFCD00002516

Formula: C₈H₆O₄
Boiling Pt: 119 - 121 °C (6 -rr)
Melting Pt: 344…348 °C (subl.)
Density: 1.540
Storage Temperature: Ambient
MDL Number: MFCD00002516
CAS Number: 121-91-5
EINECS: 204-506-4
Merck Index: 14,5197

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR