Aniline 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Aminobenzene, Phenylamine
A14443-22
A14443-0B
A14443-0I
CAAAAA14443-22EA
53.48
CAD
CAAAAA14443-22
CAAAAA14443-36
CAAAA14443-0B
CAAAA14443-0I
Aniline 99+%
Aniline
MDL: MFCD00007629
Beilstein: 605631
Notes: Miscible with alcohol, benzene chloroform.
Soluble in water
Purity: 99+%
CAS No.: 62-53-3
Molecular Formula: C6H7N
Formula Weight: 93.13
Flash Point: 70°(158°F)
Refractive Index: 1.5860
Melting Point: -6°
Boiling Point: 183-184°
Solubility: Soluble in water
Beilstein Reference: 605631
Merck Reference: 14,659
UN No.: 1547
MDL No.: MFCD00007629
Formula:
C₆H₅NH₂ MW: 93.13 g/mol Boiling Pt: 184 Melting Pt: -6.2 Density: 1.021 Flash Pt: 70 °C (158 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007629 CAS Number: 62-53-3 EINECS: 200-539-3 UN: 1547 ADR: 6.1 II Merck Index: 14,659 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...