(±)-2-Undecanol 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Undecan-2-ol, (±)-Methyl nonyl carbinol, (±)-2-Hydroxy undecane, (±)-Nonyl methyl carbinol, 2-Undecanol
A14441-09
A14441-18
CAAAA14441-09EA
112.65
CAD
CAAAA14441-09
CAAAA14441-18
CAAAA14441-30
(±)-2-Undecanol 98+%
(±)-2-Undecanol
Purity: 98+%
CAS No.: 1653-30-1
Molecular Formula: C11H24O
Formula Weight: 172.31
Flash Point: 107°(225°F)
Refractive Index: 1.4370
Odor: Mild
Melting Point: 1-3°
Boiling Point: 228-229°
Beilstein Reference: 1719795
MDL No.: MFCD00021958
Formula:
C₁₁H₂₄O MW: 172.31 g/mol Boiling Pt: 233 Melting Pt: 1…3 °C Density: 0.828 Flash Pt: 107 °C (225 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00021958 CAS Number: 1653-30-1 EINECS: 216-722-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...