4-Bromo-1,3-dimethoxybenzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Bromo-2,4-dimethoxybenzene
A14439-14
A14439-22
CAAAA14439-14EA
64.31
CAD
CAAAA14439-14
CAAAA14439-22
CAAAA14439-36
4-Bromo-1,3-dimethoxybenzene 98%
4-Bromo-1,3-dimethoxybenzene
Purity: 98%
CAS No.: 17715-69-4
Molecular Formula: C8H9BrO2
Formula Weight: 217.07
Flash Point: 113°(235°F)
Refractive Index: 1.5720
Melting Point: 24-26°
Boiling Point: 153-155°/18mm
Beilstein Reference: 1867593
MDL No.: MFCD00009844
Formula:
C₈H₉BrO₂ Boiling Pt: 153…155 °C/18 mm Melting Pt: 24…26 °C Density: 1.496 Flash Pt: 113 °C(235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009844 CAS Number: 17715-69-4 EINECS: 241-717-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...